This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213740_m1
See other RGS12 GE Assays ›
Gene Symbol:
Rgs12
Gene Name:
regulator of G-protein signaling 12
Gene Aliases:
1200016K18Rik, 4632412M04Rik, AI481290, E130309H11
Chromosome Location:
Chr.5: 34949387 - 35039642 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213740_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163512.1 NP_001156984.1 7 - 8 970 72
NM_173402.2 NP_775578.2 10 - 11 3017 72
GenBank mRNA AK004813.2 - 8 - 9 923 72
AK014569.1 - 7 - 8 972 72
AK145890.1 - 10 - 11 2966 72
AK161129.1 - 7 - 8 971 72
BC040396.1 - 10 - 11 3014 4461061 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000387.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs12

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
regulation of catalytic activity
G-protein alpha-subunit binding
receptor signaling protein activity
GTPase activator activity
protein binding
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003840.mm.1 NM_001163512.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003840.mm.1 NM_001163512.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003840.mm.1 NM_173402.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003840.mm.1 NM_173402.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003840.mm.1 TC0500000387.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0