This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213731_m1
See other NOP14 GE Assays ›
Gene Symbol:
Nop14
Gene Name:
NOP14 nucleolar protein
Gene Aliases:
2610033H07Rik, Nol14
Chromosome Location:
Chr.5: 34638536 - 34660148 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213731_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nop14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029278.2 NP_083554.2 10 - 11 1606 67
GenBank mRNA AK148605.1 - 10 - 11 1564 67
AK161728.1 - 10 - 11 1544 67
BC043043.1 - 10 - 11 1549 5701430 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nop14

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
biological_process
maturation of SSU-rRNA
ribosome biogenesis
enzyme binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0