This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213712_m1
See other HGFAC GE Assays ›
Gene Symbol:
Hgfac
Gene Name:
hepatocyte growth factor activator
Gene Aliases:
HGFA
Chromosome Location:
Chr.5: 35041514 - 35048436 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213712_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hgfac
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019447.2 NP_062320.2 4 - 5 509 107
GenBank mRNA AF099017.1 - 4 - 5 469 107
AK007987.1 - 4 - 5 507 107
AK075718.1 - 4 - 5 418 107
AK144554.1 - 4 - 5 481 107
AK149522.1 - 4 - 5 473 107
AK165433.1 - 4 - 5 486 107
AK169989.1 - 4 - 5 505 107
BC019376.1 - 4 - 5 474 5134851 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000389.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hgfac

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003866.mm.1 NM_019447.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500003866.mm.1 TC0500000389.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0