This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213333_m1
See other ZCCHC4 GE Assays ›
Gene Symbol:
Zcchc4
Gene Name:
zinc finger, CCHC domain containing 4
Gene Aliases:
4930449I23Rik
Chromosome Location:
Chr.5: 52783041 - 52824666 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213333_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zcchc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK019621.1 - 8 - 9 1022 130
AK049511.1 - 8 - 9 1029 130
AK138997.1 - 8 - 9 1041 130
BC107392.2 - 947 40057902 130
BC107393.2 - 947 40057909 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000533.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500004904.mm.1 NM_030185.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500004904.mm.1 TC0500000533.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500004905.mm.1 NM_030185.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500004905.mm.1 TC0500000533.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0