This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212776_g1
See other MSTO1 GE Assays ›
Gene Symbol:
Msto1
Gene Name:
misato homolog 1 (Drosophila)
Gene Aliases:
BC008103, mst
Chromosome Location:
Chr.3: 88909616 - 88913950 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212776_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msto1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144898.2 NP_659147.2 4 - 5 439 54
GenBank mRNA AK139708.1 - 418 54
AK163280.1 - 4 - 5 299 54
AK166236.1 - 4 - 5 411 54
AK167586.1 - 4 - 5 387 54
AK170160.1 - 4 - 5 422 54
BC008103.1 - 4 - 5 389 3966815 54
BC108354.1 - 4 - 5 373 5035560 54

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002328.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Msto1

Gene Ontology Categories:

Function(s) Process(es)

mitochondrion organization
mitochondrion distribution
molecular_function
GTPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300017751.mm.1 NM_144898.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017751.mm.1 NM_144898.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300017751.mm.1 TC0300002328.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0