Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017778.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017778.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017778.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017778.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017778.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017780.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017780.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017780.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017780.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017780.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017782.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017782.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017782.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017782.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017782.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017779.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017779.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017779.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017779.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017779.mm.1 |
TC0300002329.mm.1 |
MTA TC |