This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212665_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
Phactr3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
1500003N10Rik, 4930415A02Rik, H17739, Scapin1, Scapinin, mKIAA4224
Chromosome Location:
Chr.2: 178118975 - 178338492 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212665_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phactr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007154.3 NP_001007155.1 5 - 6 743 78
NM_001177789.1 NP_001171260.1 5 - 6 961 78
NM_001177790.1 NP_001171261.1 5 - 6 961 78
NM_001177791.1 NP_001171262.1 5 - 6 795 78
NM_028806.2 NP_083082.1 5 - 6 1200 78
GenBank mRNA AJ632220.1 - 5 - 6 758 78
AJ632221.1 - 5 - 6 796 78
AJ632222.1 - 5 - 6 743 78
AJ632223.1 - 4 - 5 633 78
AK005136.1 - 5 - 6 765 78
AK032242.1 - 5 - 6 967 78
AK038449.1 - 5 - 6 711 78
AK039213.1 - 5 - 6 750 78
AK135144.1 - 5 - 6 1201 78
AK220554.1 - 5 - 6 961 78
AY501387.1 - 5 - 6 907 78
BC058621.1 - 5 - 6 1116 5708186 78
BC059869.1 - 5 - 6 979 6809310 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002757.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phactr3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

biological_process
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001007154.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001007154.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001177789.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001177789.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001177790.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001177790.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001177791.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_001177791.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_028806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 NM_028806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200022521.mm.1 TC0200002757.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0