This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212439_m1
See other CD209A GE Assays ›
Gene Symbol:
Cd209a
Gene Name:
CD209a antigen
Gene Aliases:
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, SIGN-R1, SIGNR5
Chromosome Location:
Chr.8: 3743395 - 3748984 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212439_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd209a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133238.5 NP_573501.1 3 - 4 308 89
GenBank mRNA AF373408.1 - 3 - 4 255 89
AK054139.1 - 3 - 4 272 89
AK171639.1 - 3 - 4 252 89
AY049062.1 - 3 - 4 308 89
BC111095.1 - 2 - 3 356 40040036 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC111095.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd209a

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
regulation of gene expression
regulation of blood coagulation
regulation of T cell proliferation
viral entry into host cell
virus receptor activity
mannose binding
carbohydrate binding
metal ion binding
calcium-dependent protein binding
carbohydrate derivative binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800013320.mm.1 NM_133238.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800013320.mm.1 NM_133238.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800013320.mm.1 TC0800001619.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800013321.mm.1 NM_133238.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800013321.mm.1 NM_133238.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800013321.mm.1 TC0800001619.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0