This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212383_m1
See other RRAGD GE Assays ›
Gene Symbol:
Rragd
Gene Name:
Ras-related GTP binding D
Gene Aliases:
5730543C08Rik, AI467523, C030003H22Rik, D4Ertd174e
Chromosome Location:
Chr.4: 32982998 - 33022180 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212383_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rragd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027491.2 NP_081767.2 3 - 4 1101 116
GenBank mRNA AK017818.1 - 2 - 3 461 116
BC038137.1 - 3 - 4 590 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000275.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rragd

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
cellular response to starvation
regulation of autophagy
positive regulation of TOR signaling
cellular response to amino acid stimulus
nucleotide binding
GTPase activity
GTP binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001784.mm.1 NM_027491.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001784.mm.1 NM_027491.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400001784.mm.1 TC0400000275.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0