This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211722_g1
See other GTF3C2 GE Assays ›
Gene Symbol:
Gtf3c2
Gene Name:
general transcription factor IIIC, polypeptide 2, beta
Gene Aliases:
1300004C11Rik, 2610510G03Rik, AI225816, AU041069, TFIIIC110, mKIAA0011
Chromosome Location:
Chr.5: 31156005 - 31180658 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211722_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027901.2 NP_082177.2 9 - 10 1743 71
GenBank mRNA AK004895.1 - 9 - 10 1521 71
AK012095.1 - 2 - 3 340 71
AK035792.1 - 8 - 9 1659 71
AK043944.1 - 9 - 10 1510 71
AK083585.1 - 9 - 10 1745 71
AK083636.1 - 8 - 9 1498 71
AK129033.1 - 8 - 9 1493 71
AK138760.1 - 1512 71
AK149771.1 - 10 - 11 1640 71
AK151300.1 - 9 - 10 1508 71
AK152439.1 - 9 - 10 1508 71
AK159938.1 - 9 - 10 1511 71
BC026573.1 - 1 - 2 61 71
BC034369.1 - 9 - 10 1521 3994677 71
BC043100.1 - 9 - 10 1731 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003724.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf3c2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase III promoter
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019848.mm.1 NM_027901.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019848.mm.1 NM_027901.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500019848.mm.1 TC0500003724.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0