This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211637_m1
See other PGM1 GE Assays ›
Gene Symbol:
Pgm1
Gene Name:
phosphoglucomutase 1
Gene Aliases:
3230402E02Rik, Pgm-1, Pgm2
Chromosome Location:
Chr.5: 64092831 - 64128355 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211637_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025700.2 NP_079976.1 12 - 13 1662 84
GenBank mRNA AK004286.1 - 3 - 4 343 84
AK014332.1 - 12 - 13 1587 84
BC030869.1 - 12 - 13 1624 84
BC052762.1 - 12 - 13 1662 6510389 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000589.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pgm1

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycogen biosynthetic process
glucose metabolic process
galactose catabolic process
organic substance metabolic process
magnesium ion binding
phosphoglucomutase activity
phosphopentomutase activity
isomerase activity
intramolecular transferase activity, phosphotransferases
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500005215.mm.1 NM_025700.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005215.mm.1 NM_025700.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500005215.mm.1 TC0500000589.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0