This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211620_m1
See other SRP72 GE Assays ›
Gene Symbol:
Srp72
Gene Name:
signal recognition particle 72
Gene Aliases:
5730576P14Rik, 72kDa, AI132477, BC019196, C77589
Chromosome Location:
Chr.5: 76974701 - 76999935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211620_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srp72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025691.1 NP_079967.1 12 - 13 1251 62
GenBank mRNA AK133784.1 - 12 - 13 1252 62
AK164641.1 - 6 - 7 609 62
BC003434.1 - 12 - 13 1239 62
BC019196.1 - 12 - 13 1244 62
BC089462.1 - 3 - 4 261 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000734.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

SRP-dependent cotranslational protein targeting to membrane
response to drug
signal recognition particle binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500006599.mm.1 NM_025691.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500006599.mm.1 NM_025691.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500006599.mm.1 TC0500000734.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0