This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211381_m1
See other ADGRA2 GE Assays ›
Gene Symbol:
Adgra2
Gene Name:
adhesion G protein-coupled receptor A2
Gene Aliases:
3830613O22Rik, AU044632, Gpr125, Tem5-like
Chromosome Location:
Chr.5: 49959951 - 50058996 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211381_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgra2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133911.1 NP_598672.1 16 - 17 2676 77
GenBank mRNA AK028929.1 - 16 - 17 2615 77
AK131924.1 - 5 - 6 818 77
BC019649.1 - 5 - 6 803 77
BC020317.2 - 11 - 12 1809 77
BC052391.1 - 16 - 17 2676 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002386.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500021612.mm.1 NM_133911.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021612.mm.1 NM_133911.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021612.mm.1 TC0500002386.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0