This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210721_m1
See other HADHA GE Assays ›
Gene Symbol:
Hadha
Gene Name:
hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit
Gene Aliases:
C77020, Mtpa
Chromosome Location:
Chr.5: 30118304 - 30154980 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210721_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hadha
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178878.2 NP_849209.1 5 - 6 487 69
GenBank mRNA AK029017.1 - 5 - 6 488 69
AK035399.1 - 5 - 6 470 69
AK053986.1 - 5 - 6 485 69
AK150188.1 - 5 - 6 485 69
AK170478.1 - 5 - 6 488 69
AK170683.1 - 5 - 6 488 69
AK171469.1 - 5 - 6 488 69
AK178342.1 - 1 - 2 39 69
BC037009.1 - 5 - 6 475 5134605 69
BC046978.1 - 5 - 6 461 6315204 69
BC058569.1 - 5 - 6 483 6468514 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002154.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hadha

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
response to insulin
response to drug
oxidation-reduction process
fatty-acyl-CoA binding
catalytic activity
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acyltransferase activity
enoyl-CoA hydratase activity
oxidoreductase activity
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
lyase activity
protein complex binding
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019536.mm.1 NM_178878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019536.mm.1 NM_178878.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500019536.mm.1 TC0500002154.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0