Assay Details
Gene Symbol: Hadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178878.2 | NP_849209.1 | 13 - 14 | 1428 | 72 | |
GenBank mRNA | AK029017.1 | - | 13 - 14 | 1429 | 72 | |
AK150188.1 | - | 13 - 14 | 1426 | 72 | ||
AK170683.1 | - | 13 - 14 | 1429 | 72 | ||
AK178409.1 | - | 2 - 3 | 202 | 72 | ||
AK186817.1 | - | 1 - 2 | 105 | 72 | ||
BC027156.1 | - | 2 - 3 | 323 | 72 | ||
BC046978.1 | - | 13 - 14 | 1402 | 6315204 | 72 | |
BC058569.1 | - | 13 - 14 | 1424 | 6468514 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
