This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210380_m1
See other TRPM3 GE Assays ›
Gene Symbol:
Trpm3
Gene Name:
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases:
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
Chromosome Location:
Chr.19: 22138181 - 22989897 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210380_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trpm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035239.2 NP_001030316.1 5 - 6 809 81
NM_001035240.2 NP_001030317.1 5 - 6 809 81
NM_001035241.2 NP_001030318.1 5 - 6 809 81
NM_001035242.1 NP_001030319.1 5 - 6 809 81
NM_001035243.2 NP_001030320.1 5 - 6 809 81
NM_001035244.1 NP_001030321.1 5 - 6 857 81
NM_001035245.1 NP_001030322.1 5 - 6 857 81
NM_001035246.1 NP_001030323.1 5 - 6 857 81
NM_177341.4 NP_796315.2 5 - 6 876 81
GenBank mRNA AJ544532.1 - 5 - 6 809 81
AJ544533.1 - 5 - 6 809 81
AJ544534.1 - 5 - 6 809 81
AJ544535.1 - 5 - 6 809 81
AJ544536.1 - 5 - 6 809 81
AK044454.1 - 5 - 6 878 81
AK044533.1 - 5 - 6 655 81
AK046908.1 - 5 - 6 690 81
AK051934.1 - 5 - 6 655 81
AK052064.1 - 5 - 6 663 81
AK053673.1 - 5 - 6 694 81
AK077331.1 - 5 - 6 689 81
AK082141.1 - 5 - 6 691 81
AK084402.1 - 5 - 6 662 81
AK142504.1 - 5 - 6 690 81
AK161909.1 - 5 - 6 690 81
AK173218.1 - 5 - 6 880 81
BC037449.1 - 5 - 6 705 81
BC141400.1 - 5 - 6 852 9056013 81
BC141412.1 - 5 - 6 852 9056025 81
HM064407.1 - 5 - 6 901 81
HM064408.1 - 5 - 6 901 81
HM064409.1 - 5 - 6 901 81
HM064410.1 - 5 - 6 901 81
HM064411.1 - 5 - 6 901 81
JF706722.1 - 5 - 6 872 81
JX644970.1 - 5 - 6 901 81
JX644971.1 - 5 - 6 901 81
JX644972.1 - 5 - 6 901 81
JX644973.1 - 5 - 6 901 81
JX644974.1 - 5 - 6 901 81
JX644975.1 - 5 - 6 901 81
JX644976.1 - 5 - 6 901 81
JX644977.1 - 5 - 6 901 81
JX644978.1 - 5 - 6 901 81
JX644979.1 - 5 - 6 901 81
JX644980.1 - 5 - 6 901 81
JX644981.1 - 5 - 6 901 81
JX644982.1 - 5 - 6 901 81
JX644983.1 - 5 - 6 901 81
JX644984.1 - 5 - 6 901 81
JX644985.1 - 5 - 6 901 81
JX644986.1 - 5 - 6 901 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000356.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trpm3

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

cation transport
cation transmembrane transport
cation channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900003496.mm.1 NM_001035244.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003496.mm.1 NM_001035244.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003496.mm.1 NM_001035246.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003496.mm.1 NM_001035246.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003496.mm.1 TC1900000356.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035239.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035239.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035240.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035240.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035241.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035241.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035242.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035242.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035243.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035243.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035244.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035244.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035245.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035245.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035246.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_001035246.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_177341.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 NM_177341.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900003494.mm.1 TC1900000356.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900003498.mm.1 NM_001035246.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003498.mm.1 NM_001035246.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003498.mm.1 TC1900000356.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0