This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209966_g1
See other CCDC66 GE Assays ›
Gene Symbol:
Ccdc66
Gene Name:
coiled-coil domain containing 66
Gene Aliases:
E230015L20Rik
Chromosome Location:
Chr.14: 27481090 - 27508506 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209966_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccdc66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177111.3 NP_796085.3 12 - 13 1726 75
GenBank mRNA AK087583.1 - 4 - 5 447 75
AK170006.1 - 10 - 11 1193 75
BC070471.1 - 12 - 13 1726 30626499 75
BC072583.1 - 4 - 5 843 6403859 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001634.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccdc66

Gene Ontology Categories:

Function(s) Process(es)

retinal rod cell development
post-embryonic retina morphogenesis in camera-type eye
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400012324.mm.1 NM_177111.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400012324.mm.1 NM_177111.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400012324.mm.1 TC1400001634.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400012325.mm.1 NM_177111.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400012325.mm.1 NM_177111.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400012325.mm.1 TC1400001634.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0