This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209942_m1
See other ERC2 GE Assays ›
Gene Symbol:
Erc2
Gene Name:
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases:
6430531D06, CAST, CAST1/ERC2, D14Ertd171e, ELKS2alpha
Chromosome Location:
Chr.14: 27622410 - 28478537 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209942_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Erc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177814.4 NP_808482.2 11 - 12 2567 129
GenBank mRNA AY356531.1 - 10 - 11 2254 129
AY356533.1 - 10 - 11 2254 129
BC056760.1 - 11 - 12 2567 6331997 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000281.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Erc2

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Process(es)

protein binding
PDZ domain binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400002318.mm.1 NM_177814.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400002318.mm.1 TC1400000281.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400002317.mm.1 NM_177814.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400002317.mm.1 TC1400000281.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0