This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01208645_m1
See other FAM213A GE Assays ›
Gene Symbol:
Fam213a
Gene Name:
family with sequence similarity 213, member A
Gene Aliases:
5730469M10Rik, AU040822, Pamm
Chromosome Location:
Chr.14: 40993732 - 41013829 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01208645_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam213a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027464.3 NP_081740.2 3 - 4 334 71
GenBank mRNA AK017688.1 - 3 - 4 335 71
AK156326.1 - 3 - 4 366 71
BC056635.1 - 3 - 4 325 5064081 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001785.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam213a

Gene Ontology Categories:

Function(s) Process(es)

regulation of osteoclast differentiation
oxidation-reduction process
antioxidant activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400013735.mm.1 NM_027464.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013735.mm.1 TC1400001785.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400013736.mm.1 NM_027464.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400013736.mm.1 TC1400001785.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0