This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01208458_mH
See other ALDH1A7 GE Assays ›
Gene Symbol:
Aldh1a7
Gene Name:
aldehyde dehydrogenase family 1, subfamily A7
Gene Aliases:
AI987940, Ahd2-like, Aldh-pb, Aldh1a4, Aldhpb
Chromosome Location:
Chr.19: 20692953 - 20727556 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01208458_mH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh1a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011921.2 NP_036051.1 5 - 6 551 122
GenBank mRNA BC032880.1 - 5 - 6 528 122
BC046315.1 - 5 - 6 525 6314273 122
BC139412.1 - 5 - 6 528 8862434 122
BC139413.1 - 5 - 6 528 8862435 122
U96401.1 - 5 - 6 551 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001246.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh1a7

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
retinoic acid metabolic process
oxidation-reduction process
retinal dehydrogenase activity
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
benzaldehyde dehydrogenase (NAD+) activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900011259.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011259.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011259.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011263.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011263.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011263.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011260.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011260.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011260.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011264.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011264.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011264.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011261.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011261.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011261.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011258.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011258.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011258.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011262.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011262.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011262.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011266.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011266.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011266.mm.1 TC1900001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900011265.mm.1 NM_011921.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011265.mm.1 NM_011921.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011265.mm.1 TC1900001246.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 862870 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0