This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01208091_g1
See other RFC5 GE Assays ›
Gene Symbol:
Rfc5
Gene Name:
replication factor C (activator 1) 5
Gene Aliases:
2610020K06Rik, 2610209F07Rik, 36.5kDa, 36kDa, Recc5
Chromosome Location:
Chr.5: 117379145 - 117389023 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01208091_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rfc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028128.1 NP_082404.1 4 - 5 390 61
GenBank mRNA AK011489.1 - 4 - 5 401 61
AK150036.1 - 403 61
AK170211.1 - 4 - 5 408 61
AK171372.1 - 390 61
BC023674.1 - 3 - 4 328 61
BC089001.1 - 4 - 5 390 6415420 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA-dependent DNA replication
nucleotide binding
DNA binding
DNA clamp loader activity
ATP binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020337 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020332 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020331 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020334 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020335 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020333 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 BC023674.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 BC089001.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 AK170211.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 AK171372.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 AK150036.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 BC023674.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 BC089001.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 AK170211.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 AK171372.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246339 AK150036.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678517 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443187 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553618 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444587 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252225 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652356 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921464 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864464 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149752 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157503 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 BC023674.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 BC089001.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 AK170211.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 AK171372.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 AK150036.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 BC023674.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 BC089001.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 AK170211.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 AK171372.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246339 AK150036.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 BC023674.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 BC089001.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 AK170211.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 AK171372.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 AK150036.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 BC023674.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 BC089001.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 AK170211.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 AK171372.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246339 AK150036.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157503 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05711230 - -
Whole Mouse Genome G4122A A_51_P246339 BC023674.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 BC089001.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 AK170211.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 AK171372.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 AK150036.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 BC023674.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 BC089001.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 AK170211.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 AK171372.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P246339 AK150036.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157503 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04356734 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020334 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020332 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020337 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020331 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020335 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24020333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020204 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020194 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020311 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24020323 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04407858 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020350 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020285 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04407863 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04114962 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04850975 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04264665 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04520313 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020268 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04114961 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04264674 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04188830 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020295 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020336 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020281 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020272 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020308 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020337 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020274 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04868487 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04407844 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020348 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020340 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020300 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04608679 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04264664 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04356738 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020299 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04608680 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04264673 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020354 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020333 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04188835 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020301 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020273 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020306 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020279 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020293 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04114965 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04850976 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020291 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020287 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020329 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04900423 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04188829 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020334 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04114963 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020315 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04467945 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24020316 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04927828 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04225332 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04467943 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10533091 - -
Affymetrix Mouse Gene 1.0 ST Array 10533093 - -
Affymetrix Mouse Gene 1.0 ST Array 10533092 - -
Affymetrix Mouse Gene 1.0 ST Array 10533094 - -
Affymetrix HT MG-430 PM Array Plate 1452917_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10533090 - -
Mouse Genome 430 2.0 Array 1452917_PM_at - -
Mouse Genome 430 2.0 Array 1452917_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72151_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72151_at - -
Affymetrix Mouse Clariom S Array PSR0500027862.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027862.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027862.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027859.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027859.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027859.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027860.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027860.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027860.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027861.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027861.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027861.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027850.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027850.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027850.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027854.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027854.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027854.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027857.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027857.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027857.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027848.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027848.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027848.mm.1 TC0500003131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027849.mm.1 NM_028128.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027849.mm.1 NM_028128.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027849.mm.1 TC0500003131.mm.1 MTA TC
Mouse Expression Set 430 1452917_PM_at - -
Mouse Expression Set 430 1452917_at - -
Murine Genome U74v2 Set 104001_PM_at - -
Murine Genome U74v2 Set 95612_PM_at - -
Murine Genome U74v2 Set 95612_at - -
Murine Genome U74v2 Set 104001_at - -
Mouse Genome 430A 2.0 Array 1452917_at - -
Mouse Genome 430A 2.0 Array 1452917_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1452917_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1452917_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3800452 - -
MouseWG-6 v2 Expression BeadChip 6020091 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662657 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2637203 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2662657 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2637203 - -
MouseRef-8 v2 Expression BeadChip ILMN_2637203 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0