This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01207777_g1
See other HNRNPDL GE Assays ›
Gene Symbol:
Hnrnpdl
Gene Name:
heterogeneous nuclear ribonucleoprotein D-like
Gene Aliases:
AA407431, AA959857, D5Ertd650e, D5Wsu145e, Hnrnpdl1, Hnrpdl, JKTBP, hnRNP DL, hnRNP-DL
Chromosome Location:
Chr.5: 100033577 - 100039950 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01207777_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hnrnpdl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016690.4 NP_057899.2 4 - 5 968 92
GenBank mRNA AB017020.1 - 4 - 5 674 92
AK172629.1 - 4 - 5 627 92
BC021374.1 - 4 - 5 626 5039077 92
CX219512.1 - 3 - 4 343 92

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hnrnpdl

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of gene expression
mRNA metabolic process
nucleotide binding
nucleic acid binding
DNA binding
double-stranded DNA binding
single-stranded DNA binding
RNA binding
poly(A) binding
poly(G) binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500024882.mm.1 NM_016690.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500024882.mm.1 NM_016690.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500024882.mm.1 TC0500002828.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500024881.mm.1 NM_016690.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500024881.mm.1 NM_016690.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500024881.mm.1 TC0500002828.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0