This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01207494_m1
See other TRIM56 GE Assays ›
Gene Symbol:
Trim56
Gene Name:
tripartite motif-containing 56
Gene Aliases:
A130009K11Rik, Gm452, RNF109
Chromosome Location:
Chr.5: 137111286 - 137116466 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01207494_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trim56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201373.4 NP_958761.1 1 - 2 94 98
GenBank mRNA AK037350.1 - 1 - 2 95 98
BC045615.1 - 1 - 2 84 4976515 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK037350.1, BC045615.1, NM_201373.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trim56

Gene Ontology Categories:

Function(s) Process(es)

immune system process
interferon-beta production
response to type I interferon
innate immune response
negative regulation of viral entry into host cell
defense response to virus
protein K63-linked ubiquitination
negative regulation of viral release from host cell
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
poly(A) RNA binding
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139763 BC045615.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724708 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346584 BC039782.1 genbank_mrna_mouse
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106283 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139763 BC045615.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244820 BC045615.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244820 AK037350.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244820 NM_201373.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124658 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124648 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124651 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124660 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124680 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124650 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124646 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124661 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124668 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124655 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124662 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124656 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124666 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124674 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124670 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124682 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124672 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124665 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124653 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124676 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124645 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05759854 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106283 - -
Whole Mouse Genome G4122A A_51_P244820 BC045615.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P244820 AK037350.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P244820 NM_201373.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129681 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112484 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106283 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04593667 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04321859 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04593665 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04321860 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04593668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124593 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04321804 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124614 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124586 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124591 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124567 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124643 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124568 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124626 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04497969 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124589 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124580 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124666 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124579 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04392916 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124622 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124653 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04668643 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04904734 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124565 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124585 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124684 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124655 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04679995 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124663 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124676 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04672990 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124672 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04679997 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124683 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124629 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124575 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04399426 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124639 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124668 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124618 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124642 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124578 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124665 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124603 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124661 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04679994 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124691 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124671 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04575903 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124633 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124674 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124632 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124657 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124611 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124688 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124617 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124625 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124572 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124636 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124658 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04679998 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04672973 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04668641 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124689 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124680 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124690 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124571 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04668640 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124620 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124678 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124630 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124607 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124648 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124577 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04672975 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124646 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124685 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04123614 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04678755 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24124660 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10534682 - -
Affymetrix Mouse Gene 1.0 ST Array 10534681 - -
Affymetrix Mouse Gene 1.0 ST Array 10534683 - -
Affymetrix Mouse Gene 1.0 ST Array 10534680 - -
Affymetrix Mouse Gene 1.1 ST Array 10534679 - -
Affymetrix Mouse Clariom S Array PSR0500031117.mm.1 NM_201373.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031117.mm.1 NM_201373.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500031117.mm.1 TC0500003433.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500031118.mm.1 NM_201373.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031118.mm.1 NM_201373.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500031118.mm.1 TC0500003433.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500031119.mm.1 NM_201373.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031119.mm.1 NM_201373.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500031119.mm.1 TC0500003433.mm.1 MTA TC
Murine Genome U74v2 Set 131166_at - -
Murine Genome U74v2 Set 131166_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 687527 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1580528 - -
Illumina mouse-6 v1.0 expression beadchip 1580528 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001580528 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2441921 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2441921 - -
mouse-6 v1.1 (Illumina) scl0384309.1_171-S - -
Illumina Sentrix mouse-6.v1 bead array 1580528 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0384309.1_171-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580528 - -
MouseRef-8 v2 Expression BeadChip ILMN_2441921 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2441921 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0