This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01207193_m1
See other TMEM132D GE Assays ›
Gene Symbol:
Tmem132d
Gene Name:
transmembrane protein 132D
Gene Aliases:
C630028F04Rik
Chromosome Location:
Chr.5: 127781629 - 128433077 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01207193_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem132d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172885.2 NP_766473.1 6 - 7 2361 61
GenBank mRNA AB100355.1 - 6 - 7 1815 61
AK083210.1 - 6 - 7 2362 61
BC140953.1 - 6 - 7 1901 9055565 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003286.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem132d

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500029753.mm.1 NM_172885.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500029753.mm.1 NM_172885.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500029753.mm.1 TC0500003286.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0