This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01206158_m1
See other EFR3A GE Assays ›
Gene Symbol:
Efr3a
Gene Name:
EFR3 homolog A (S. cerevisiae)
Gene Aliases:
A130089M23Rik, BB071175, C76891, C920006C10Rik, D030063F01Rik, mKIAA0143
Chromosome Location:
Chr.15: 65787020 - 65873812 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01206158_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Efr3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133766.3 NP_598527.2 19 - 20 2352 67
GenBank mRNA AB158474.1 - 19 - 20 2356 67
AK018695.1 - 14 - 15 1567 67
AK030074.1 - 19 - 20 2353 67
AK032461.1 - 19 - 20 2325 67
AK044000.1 - 19 - 20 2346 67
AK044475.1 - 19 - 20 2348 67
AK129065.1 - 19 - 20 2330 67
AK161250.1 - 19 - 20 2337 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Efr3a

Gene Ontology Categories:

Function(s) Process(es)

single organismal cell-cell adhesion
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0