This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01206130_m1
See other PCSK5 GE Assays ›
Gene Symbol:
Pcsk5
Gene Name:
proprotein convertase subtilisin/kexin type 5
Gene Aliases:
PC5, PC5/6A, PC5A, PC6, SPC6, b2b1549Clo, b2b585Clo
Chromosome Location:
Chr.19: 17432832 - 17837632 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01206130_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcsk5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163144.1 NP_001156616.1 11 - 12 1912 63
NM_001190483.1 NP_001177412.1 11 - 12 1912 63
GenBank mRNA AK143998.1 - 10 - 11 1809 63
BC013068.1 - 11 - 12 1532 63
D12619.1 - 11 - 12 1501 63
D17583.1 - 4 - 5 445 63
L14932.1 - 11 - 12 1462 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001219.mm.1

Set Membership:

Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

kidney development
signal peptide processing
proteolysis
heart development
female pregnancy
embryo implantation
anterior/posterior pattern specification
protein processing
peptide hormone processing
viral life cycle
respiratory tube development
limb morphogenesis
cytokine biosynthetic process
peptide biosynthetic process
embryonic digestive tract development
embryonic skeletal system development
endopeptidase activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
peptide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900011129.mm.1 NM_001163144.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011129.mm.1 NM_001163144.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011129.mm.1 NM_001190483.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900011129.mm.1 TC1900001219.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0