This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01206049_m1
See other SLC25A24 GE Assays ›
Gene Symbol:
Slc25a24
Gene Name:
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 24
Gene Aliases:
2610016M12Rik
Chromosome Location:
Chr.3: 109123149 - 109168409 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01206049_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172685.3 NP_766273.1 2 - 3 368 136
GenBank mRNA AK032806.1 - 2 - 3 369 136
AK146034.1 - 2 - 3 367 136
AK167778.1 - 2 - 3 359 136
AK168090.1 - 2 - 3 359 136
AK170598.1 - 2 - 3 360 136
AK187917.1 - 1 - 2 137 136
BC055369.1 - 2 - 3 357 30055307 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001159.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a24

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
mitochondrial transport
regulation of cell death
purine nucleoside transmembrane transport
ATP transport
cellular response to oxidative stress
transmembrane transport
cellular response to calcium ion
anion transmembrane transport
nucleotide transmembrane transport
ATP transmembrane transporter activity
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300009487.mm.1 NM_172685.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009487.mm.1 NM_172685.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300009487.mm.1 TC0300001159.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0