This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205889_m1
See other VSIG10L GE Assays ›
Gene Symbol:
Vsig10l
Gene Name:
ZV-set and immunoglobulin domain containing 10 like
Gene Aliases:
-
Chromosome Location:
Chr.7: 43463061 - 43472023 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205889_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vsig10l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290316.1 NP_001277245.1 5 - 6 1389 80
GenBank mRNA BC112410.1 - 5 - 6 1383 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171236 BC112410.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120777 AK008908.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120777 AK008908.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171236 BC112410.1 mouse_transcriptome
Agilent Six_ChIP_02_of_03_v2 A_68_P06197943 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082521 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082530 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082544 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06321971 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082526 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082522 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082531 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25082525 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10552428 - -
Affymetrix Mouse Gene 1.0 ST Array 10552429 - -
Affymetrix Mouse Gene 1.0 ST Array 10552430 - -
Affymetrix Mouse Gene 1.0 ST Array 10552427 - -
Affymetrix Mouse Gene 1.0 ST Array 10552431 - -
Affymetrix Mouse Gene 1.0 ST Array 10552426 - -
Affymetrix Mouse Gene 1.0 ST Array 10552432 - -
Affymetrix HT MG-430 PM Array Plate 1445472_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10552425 - -
Mouse Genome 430 2.0 Array 1445472_PM_at - -
Mouse Genome 430 2.0 Array 1445472_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 75690_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 75690_at - -
Mouse Expression Set 430 1445472_PM_at - -
Mouse Expression Set 430 1445472_at - -
Affymetrix HT Mouse Genome 430B Array 1445472_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1445472_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 3190450 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003190450 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213261 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1213261 - -
mouse-6 v1.1 (Illumina) scl075690.1_185-S - -
Illumina Sentrix mouse-6.v1 bead array 3190450 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl075690.1_185-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190450 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213261 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0