This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205826_g1
See other SMPD5 GE Assays ›
Gene Symbol:
Smpd5
Gene Name:
sphingomyelin phosphodiesterase 5
Gene Aliases:
ENSMUSG00000071724, Gm10345, MA-nSMase
Chromosome Location:
Chr.15: 76293869 - 76296896 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205826_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smpd5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195537.1 NP_001182466.1 3 - 4 1073 66
GenBank mRNA AK005822.1 - 2 - 3 355 66
BY705974.1 - 2 - 3 352 66
CF197814.1 - 1 - 2 149 66
GU144514.1 - 3 - 4 1073 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000583.mm.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ceramide metabolic process
sphingomyelin phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004020.mm.1 NM_001195537.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004020.mm.1 NM_001195537.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004020.mm.1 TC1500000583.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500004022.mm.1 NM_001195537.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004022.mm.1 NM_001195537.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004022.mm.1 TC1500000583.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0