This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205822_g1
See other PGAM5 GE Assays ›
Gene Symbol:
Pgam5
Gene Name:
phosphoglycerate mutase family member 5
Gene Aliases:
2610528A17Rik
Chromosome Location:
Chr.5: 110259135 - 110269899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205822_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgam5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163538.1 NP_001157010.1 4 - 5 641 66
GenBank mRNA AK012159.1 - 4 - 5 617 66
AK049246.1 - 4 - 5 601 66
AK146216.1 - 4 - 5 642 66
AK169643.1 - 4 - 5 604 66
BC021317.1 - 4 - 5 591 66
BC052179.1 - 4 - 5 565 66
BC145467.1 - 4 - 5 624 9054029 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002985.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

isomerase mutase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

programmed cell death
dephosphorylation
intracellular signal transduction
positive regulation of GTPase activity
necroptotic process
phosphoprotein phosphatase activity
GTPase activator activity
receptor signaling protein serine/threonine phosphatase activity
hydrolase activity
phosphatase activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500026398.mm.1 NM_001163538.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026398.mm.1 NM_001163538.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026398.mm.1 NM_028273.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026398.mm.1 NM_028273.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500026398.mm.1 TC0500002985.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0