This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205807_m1
See other ANKLE2 GE Assays ›
Gene Symbol:
Ankle2
Gene Name:
ankyrin repeat and LEM domain containing 2
Gene Aliases:
1110001J12Rik, AI661024, D5Ertd585e
Chromosome Location:
Chr.5: 110231004 - 110256651 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205807_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ankle2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253814.1 NP_001240743.1 3 - 4 1105 143
GenBank mRNA AK043925.1 - 3 - 4 946 143
AK129192.1 - 2 - 3 431 143
AK132187.1 - 3 - 4 1107 143
AK142497.1 - 1 - 2 154 143

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
mitotic nuclear envelope reassembly
positive regulation of protein dephosphorylation
negative regulation of phosphorylation
cell division
protein phosphatase 2A binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0