Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500009713.mm.1 |
NM_001253814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009713.mm.1 |
NM_001253814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009713.mm.1 |
NM_027922.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009713.mm.1 |
NM_027922.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009713.mm.1 |
TC0500001136.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009712.mm.1 |
NM_001253814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009712.mm.1 |
NM_001253814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009712.mm.1 |
NM_027922.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009712.mm.1 |
NM_027922.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009712.mm.1 |
TC0500001136.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009714.mm.1 |
NM_001253814.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009714.mm.1 |
NM_001253814.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009714.mm.1 |
NM_027922.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009714.mm.1 |
NM_027922.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009714.mm.1 |
TC0500001136.mm.1 |
MTA TC |