This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205712_g1
See other ADCK5 GE Assays ›
Gene Symbol:
Adck5
Gene Name:
aarF domain containing kinase 5
Gene Aliases:
A230108P17
Chromosome Location:
Chr.15: 76576359 - 76595812 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205712_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adck5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172960.3 NP_766548.2 4 - 5 468 74
GenBank mRNA AK163557.1 - 4 - 5 416 74
BC030881.1 - 4 - 5 448 74
BQ886383.1 - 4 - 5 391 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000599.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adck5

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
phosphorylation
molecular_function
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
3-phosphoinositide-dependent protein kinase activity
DNA-dependent protein kinase activity
AMP-activated protein kinase activity
cyclic nucleotide-dependent protein kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
calcium-dependent protein kinase C activity
receptor signaling protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
JUN kinase activity
JUN kinase kinase kinase activity
MAP kinase kinase activity
MAP kinase kinase kinase activity
ribosomal protein S6 kinase activity
MAP kinase kinase kinase kinase activity
JUN kinase kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
SAP kinase activity
transmembrane receptor protein kinase activity
cyclin-dependent protein kinase activating kinase activity
GTP-dependent protein kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004305.mm.1 NM_172960.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004305.mm.1 NM_172960.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500004305.mm.1 NR_073123.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004305.mm.1 TC1500000599.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0