Assay Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 22 - 23 | 2703 | 116 | |
NM_053193.2 | NP_444423.1 | 22 - 23 | 2703 | 116 | ||
GenBank mRNA | AF322193.1 | - | 22 - 23 | 2674 | 116 | |
AK149914.1 | - | 15 - 16 | 1702 | 116 | ||
AK155806.1 | - | 22 - 23 | 2625 | 116 | ||
BC011458.1 | - | 18 - 19 | 2139 | 116 | ||
BC049941.1 | - | 22 - 23 | 2525 | 116 | ||
BC056388.1 | - | 22 - 23 | 2703 | 6854851 | 116 | |
BC057945.1 | - | 7 - 8 | 1034 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
