This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205549_m1
See other BAX GE Assays ›
Gene Symbol:
Bax
Gene Name:
BCL2-associated X protein
Gene Aliases:
-
Chromosome Location:
Chr.7: 45461695 - 45466903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205549_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bax
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007527.3 NP_031553.1 3 - 4 360 89
GenBank mRNA AK013298.1 - 3 - 4 270 89
AK149994.1 - 3 - 4 253 89
AK159233.1 - 3 - 4 325 89
AY095934.1 - 2 - 3 707 89
BC018228.1 - 3 - 4 305 3968903 89
BC053380.1 - 3 - 4 277 6489475 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003007.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bax

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
neuron migration
leukocyte homeostasis
T cell homeostatic proliferation
B cell homeostasis
B cell apoptotic process
kidney development
release of cytochrome c from mitochondria
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
blood vessel remodeling
myeloid cell homeostasis
B cell negative selection
B cell homeostatic proliferation
positive regulation of B cell apoptotic process
glycosphingolipid metabolic process
regulation of nitrogen utilization
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
obsolete transformed cell apoptotic process
cellular response to DNA damage stimulus
inner mitochondrial membrane organization
outer mitochondrial membrane organization
germ cell development
spermatogenesis
nervous system development
sex differentiation
mitochondrial fusion
cell proliferation
negative regulation of cell proliferation
male gonad development
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
apoptotic mitochondrial changes
fertilization
response to wounding
response to toxic substance
response to salt stress
post-embryonic development
response to ionizing radiation
establishment or maintenance of transmembrane electrochemical gradient
response to gamma radiation
positive regulation of calcium ion transport into cytosol
hypothalamus development
cerebral cortex development
negative regulation of protein binding
positive regulation of protein oligomerization
negative regulation of endoplasmic reticulum calcium ion concentration
release of matrix enzymes from mitochondria
negative regulation of peptidyl-serine phosphorylation
regulation of mammary gland epithelial cell proliferation
cellular response to UV
limb morphogenesis
ectopic germ cell programmed cell death
odontogenesis of dentin-containing tooth
regulation of apoptotic process
positive regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
mitochondrial fragmentation involved in apoptotic process
development of secondary sexual characteristics
cellular respiration
retinal cell programmed cell death
positive regulation of developmental pigmentation
negative regulation of fibroblast proliferation
spermatid differentiation
post-embryonic camera-type eye morphogenesis
response to axon injury
homeostasis of number of cells
homeostasis of number of cells within a tissue
protein oligomerization
protein homooligomerization
positive regulation of release of sequestered calcium ion into cytosol
neuron apoptotic process
regulation of cell cycle
regulation of mitochondrial membrane potential
transmembrane transport
Sertoli cell proliferation
retina development in camera-type eye
positive regulation of apoptotic process involved in mammary gland involution
vagina development
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
thymocyte apoptotic process
mitochondrion morphogenesis
cellular response to organic substance
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
intrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum unfolded protein response
regulation of mitochondrial membrane permeability involved in apoptotic process
positive regulation of mitochondrial membrane permeability involved in apoptotic process
apoptotic process involved in patterning of blood vessels
apoptotic process involved in embryonic digit morphogenesis
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
positive regulation of apoptotic DNA fragmentation
retinal cell apoptotic process
B cell receptor apoptotic signaling pathway
negative regulation of apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of intrinsic apoptotic signaling pathway
protein binding
lipid binding
channel activity
heat shock protein binding
protein complex binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
chaperone binding
BH domain binding
BH3 domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700027236.mm.1 NM_007527.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700027236.mm.1 NM_007527.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700027236.mm.1 TC0700003007.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0