This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204647_m1
See other SSH1 GE Assays ›
Gene Symbol:
Ssh1
Gene Name:
slingshot homolog 1 (Drosophila)
Gene Aliases:
AW551225, Gm1394, Gm1395, SSH-1, mSSH-1L
Chromosome Location:
Chr.5: 113937096 - 113993757 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204647_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198109.4 NP_932777.2 8 - 9 760 121
GenBank mRNA AB099287.1 - 8 - 9 795 121
AK089308.1 - 8 - 9 764 121
AK155557.1 - 8 - 9 785 121
AK165380.1 - 7 - 8 725 121
AK170787.1 - 7 - 8 685 121
AK171210.1 - 8 - 9 809 121
AK171556.1 - 8 - 9 775 121
AK173144.1 - 7 - 8 716 121
BC046529.1 - 2 - 3 242 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003054.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssh1

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
protein dephosphorylation
regulation of actin polymerization or depolymerization
regulation of lamellipodium assembly
dephosphorylation
actin cytoskeleton organization
regulation of cellular protein metabolic process
regulation of axonogenesis
cellular response to ATP
DNA binding
actin binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500027152.mm.1 NM_198109.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027152.mm.1 NM_198109.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500027152.mm.1 TC0500003054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0