This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204563_g1
See other RAD21 GE Assays ›
Gene Symbol:
Rad21
Gene Name:
RAD21 homolog (S. pombe)
Gene Aliases:
SCC1, mKIAA0078
Chromosome Location:
Chr.15: 51962604 - 51993538 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204563_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rad21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009009.4 NP_033035.3 6 - 7 926 139
GenBank mRNA AF332085.1 - 5 - 6 681 139
AF332086.1 - 5 - 6 681 139
AK128984.1 - 6 - 7 815 139
AK139492.1 - 5 - 6 579 139
AK145591.1 - 6 - 7 893 139
AK163992.1 - 787 139
AK168718.1 - 6 - 7 914 139
AK168896.1 - 6 - 7 875 139
BC043032.1 - 5 - 6 695 5696540 139
BC129916.1 - 5 - 6 725 40111820 139
BC129917.1 - 5 - 6 726 40111821 139
D37790.1 - 5 - 6 613 139
X98293.1 - 5 - 6 681 139

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rad21

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
cell division
protein localization to chromatin
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500011441.mm.1 NM_009009.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011441.mm.1 NM_009009.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500011441.mm.1 TC1500001506.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0