This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204438_m1
See other TJP2 GE Assays ›
Gene Symbol:
Tjp2
Gene Name:
tight junction protein 2
Gene Aliases:
ZO-2
Chromosome Location:
Chr.19: 24094502 - 24225128 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204438_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tjp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198985.1 NP_001185914.1 9 - 10 1587 90
NM_011597.4 NP_035727.2 9 - 10 1577 90
GenBank mRNA AF113005.1 - 9 - 10 1435 90
AK028494.1 - 9 - 10 1506 90
BC034677.1 - 9 - 10 1495 3155078 90
BC039924.1 - 9 - 10 1506 3155079 90
BC042695.1 - 9 - 10 1479 90
CN527767.1 - 1 - 2 120 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001273.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tjp2

Panther Classification:

Molecular Function -

tight junction

Gene Ontology Categories:

Function(s) Process(es)

intestinal absorption
establishment of endothelial intestinal barrier
regulation of membrane permeability
protein binding
protein C-terminus binding
protein domain specific binding
protein binding, bridging

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900011516.mm.1 NM_001198985.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011516.mm.1 NM_001198985.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011516.mm.1 NM_011597.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900011516.mm.1 NM_011597.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900011516.mm.1 TC1900001273.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0