This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204202_g1
See other SDHAF2 GE Assays ›
Gene Symbol:
Sdhaf2
Gene Name:
succinate dehydrogenase complex assembly factor 2
Gene Aliases:
0610038F07Rik, AA407634, AW049997
Chromosome Location:
Chr.19: 10500512 - 10525209 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204202_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdhaf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025333.4 NP_079609.2 2 - 3 366 112
GenBank mRNA AK013454.1 - 2 - 3 271 112
AK075594.1 - 2 - 3 271 112
AK160957.1 - 2 - 3 273 112
AK167228.1 - 2 - 3 302 112
BC029630.1 - 1 - 2 85 112
BC058955.1 - 2 - 3 315 112
BC060090.1 - 2 - 3 315 5698361 112
BC085277.1 - 2 - 3 268 30441476 112

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdhaf2

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, succinate to ubiquinone
protein dephosphorylation
negative regulation of epithelial to mesenchymal transition
protein-FAD linkage
negative regulation of canonical Wnt signaling pathway
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0