Assay Details
| Gene Symbol: Ly6k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014121.1 | - | 389 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D730001G18 gene | D730001G18Rik | -19.7kb | Anti-sense | Chr. 15 - 74770908 - 74778859 on NCBI Build GRCm38 | |
| Downstream | GPI anchored molecule like protein | Gml |
EG625599, HemT |
13.5kb | Anti-sense | Chr. 15 - 74813435 - 74818815 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ly6k
Genomic Map