This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01203999_g1
See other VWCE GE Assays ›
Gene Symbol:
Vwce
Gene Name:
von Willebrand factor C and EGF domains
Gene Aliases:
1300015B04Rik
Chromosome Location:
Chr.19: 10634229 - 10665210 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01203999_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vwce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027913.1 NP_082189.1 18 - 19 2357 57
GenBank mRNA AK005010.1 - 18 - 19 2338 57
AK153937.1 - 18 - 19 2137 57
BC043473.1 - 18 - 19 2277 5346280 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000206.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
positive regulation of cell proliferation
regulation of signal transduction
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002615.mm.1 NM_027913.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002615.mm.1 NM_027913.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002615.mm.1 TC1900000206.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0