This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01203407_m1
See other ADGRD1 GE Assays ›
Gene Symbol:
Adgrd1
Gene Name:
adhesion G protein-coupled receptor D1
Gene Aliases:
E230012M21Rik, Gpr133
Chromosome Location:
Chr.5: 129096666 - 129204599 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01203407_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081342.1 NP_001074811.1 17 - 18 2266 87
GenBank mRNA AK054021.1 - 2 - 3 113 87
AK153762.1 - 17 - 18 2269 88
AY255618.1 - 2 - 3 151 87
BC151161.1 - 17 - 18 1968 9088050 87
BC157994.1 - 16 - 17 1872 9007307 87
BC158001.1 - 16 - 17 1872 9007310 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK054021.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013516.mm.1 NM_001081342.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013516.mm.1 NM_001081342.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013516.mm.1 TC0500001501.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500013515.mm.1 NM_001081342.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013515.mm.1 NM_001081342.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013515.mm.1 TC0500001501.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0