This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01203405_m1
See other ADGRD1 GE Assays ›
Gene Symbol:
Adgrd1
Gene Name:
adhesion G protein-coupled receptor D1
Gene Aliases:
E230012M21Rik, Gpr133
Chromosome Location:
Chr.5: 129096666 - 129204599 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01203405_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081342.1 NP_001074811.1 15 - 16 2042 126
GenBank mRNA AK153762.1 - 15 - 16 2044 126
BC151161.1 - 15 - 16 1744 9088050 126
BC157994.1 - 14 - 15 1648 9007307 126
BC158001.1 - 14 - 15 1648 9007310 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001501.mm.1

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013513.mm.1 NM_001081342.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013513.mm.1 NM_001081342.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013513.mm.1 TC0500001501.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0