This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202697_m1
See other GPN3 GE Assays ›
Gene Symbol:
Gpn3
Gene Name:
GPN-loop GTPase 3
Gene Aliases:
A930018B01Rik, Atpbd1c, D5Ertd708e
Chromosome Location:
Chr.5: 122372508 - 122382771 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202697_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpn3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024216.1 NP_077178.1 4 - 5 568 78
GenBank mRNA AK016996.1 - 4 - 5 499 78
AK042405.1 - 1191 78
AK080720.1 - 4 - 5 515 78
AK087968.1 - 4 - 5 519 78
AK146783.1 - 4 - 5 506 78
BC003341.1 - 4 - 5 568 3257412 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001390.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpn3

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
GTP binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500012292.mm.1 NM_024216.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500012292.mm.1 NM_024216.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500012292.mm.1 TC0500001390.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0