This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201778_m1
See other SSC4D GE Assays ›
Gene Symbol:
Ssc4d
Gene Name:
scavenger receptor cysteine rich family, 4 domains
Gene Aliases:
C330016E03Rik, S4D-SRCRB, SRCRB-S4D, Srcrb4d
Chromosome Location:
Chr.5: 135960220 - 135974535 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201778_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssc4d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160366.1 NP_001153838.1 10 - 11 1737 87
GenBank mRNA AK082786.1 - 2 - 3 148 87
BC117747.1 - 7 - 8 1244 87
BC126947.1 - 7 - 8 1244 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003411.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

biological_process
scavenger receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016227 BC117747.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016227 BC126947.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016227 NM_001160366.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016227 AK082786.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016222 AK082786.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 152050 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956354 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750556 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926061 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158069 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596189 AK016764.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503722 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507569 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852960 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61655 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135715 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016227 BC117747.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016227 BC126947.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016227 NM_001160366.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016227 AK082786.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016222 AK082786.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05756887 - -
Whole Mouse Genome G4122A A_51_P165408 BC117747.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P165408 BC126947.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P165408 NM_001160366.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P165408 AK082786.1 mouse_transcriptome
Agilent Six_ChIP_01_of_03_v2 A_68_P04704454 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118696 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118767 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118698 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04848472 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118715 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118742 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118776 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118772 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118730 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04856947 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118726 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04149936 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118697 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04451174 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118744 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04385564 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04451182 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118765 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118723 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118718 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118703 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118771 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04767717 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118735 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118731 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118719 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118713 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04385571 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118739 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118708 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04124612 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04385577 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118773 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118732 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04767715 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118740 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04574584 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04451172 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118720 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04121403 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118712 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04385578 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04215908 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24118707 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10534538 - -
Affymetrix Mouse Gene 1.0 ST Array 10534543 - -
Affymetrix Mouse Gene 1.0 ST Array 10534548 - -
Affymetrix Mouse Gene 1.0 ST Array 10534539 - -
Affymetrix Mouse Gene 1.0 ST Array 10534547 - -
Affymetrix Mouse Gene 1.0 ST Array 10534544 - -
Affymetrix Mouse Gene 1.0 ST Array 10534542 - -
Affymetrix Mouse Gene 1.0 ST Array 10534546 - -
Affymetrix Mouse Gene 1.0 ST Array 10534541 - -
Affymetrix Mouse Gene 1.0 ST Array 10534545 - -
Affymetrix HT MG-430 PM Array Plate 1438147_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10534537 - -
Mouse Genome 430 2.0 Array 1438147_at - -
Mouse Genome 430 2.0 Array 1438147_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 109267_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 109267_at - -
Affymetrix Mouse Clariom S Array PSR0500030814.mm.1 NM_001160366.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030814.mm.1 NM_001160366.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030814.mm.1 TC0500003411.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030815.mm.1 NM_001160366.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030815.mm.1 NM_001160366.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030815.mm.1 TC0500003411.mm.1 MTA TC
Mouse Expression Set 430 1438147_PM_at - -
Mouse Expression Set 430 1438147_at - -
Murine Genome U74v2 Set 95307_PM_at - -
Murine Genome U74v2 Set 95307_at - -
Affymetrix HT Mouse Genome 430B Array 1438147_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1438147_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 921535 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1690446 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2654947 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2654947 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0