This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201317_m1
See other HNRNPD GE Assays ›
Gene Symbol:
Hnrnpd
Gene Name:
heterogeneous nuclear ribonucleoprotein D
Gene Aliases:
Auf1, Hnrpd
Chromosome Location:
Chr.5: 99955935 - 99979015 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201317_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hnrnpd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077265.2 NP_001070733.1 7 - 8 1315 108
NM_001077266.2 NP_001070734.1 6 - 7 1258 108
GenBank mRNA AK053251.1 - 1 - 2 99 108
BC011172.1 - 7 - 8 990 108
CA451481.1 - 5 - 6 596 108
CX567003.1 - 3 - 4 290 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of circadian rhythm
regulation of mRNA stability
positive regulation of translation
mRNA stabilization
rhythmic process
circadian regulation of translation
nucleotide binding
nucleic acid binding
DNA binding
RNA binding
protein binding
telomeric DNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0