This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201080_m1
See other STAMBPL1 GE Assays ›
Gene Symbol:
Stambpl1
Gene Name:
STAM binding protein like 1
Gene Aliases:
1700095N21Rik, 8230401J17Rik, ALMalpha, AMSH-FP
Chromosome Location:
Chr.19: 34192190 - 34240657 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201080_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stambpl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029682.4 NP_083958.3 4 - 5 739 77
GenBank mRNA AB010121.1 - 4 - 5 434 77
AB010122.1 - 4 - 5 398 77
AB066211.1 - 4 - 5 739 77
AB066213.1 - 4 - 5 422 77
AK085665.1 - 3 - 4 3130 77
AK138126.1 - 4 - 5 470 77
BC018343.1 - 4 - 5 465 4010423 77
BC023097.1 - 4 - 5 708 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000495.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stambpl1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
biological_process
molecular_function
peptidase activity
metallopeptidase activity
hydrolase activity
pyroglutamyl-peptidase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004421.mm.1 NM_029682.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004421.mm.1 NM_029682.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900004421.mm.1 TC1900000495.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0