This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200531_g1
See other GPAA1 GE Assays ›
Gene Symbol:
Gpaa1
Gene Name:
GPI anchor attachment protein 1
Gene Aliases:
C80044, mGAA1
Chromosome Location:
Chr.15: 76331294 - 76334900 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200531_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpaa1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010331.2 NP_034461.1 9 - 10 1361 81
GenBank mRNA AB002136.1 - 9 - 10 1290 81
AB006970.1 - 9 - 10 1297 81
AK077518.1 - 9 - 10 1361 81
AK164219.1 - 9 - 10 1307 81
AK164978.1 - 9 - 10 1361 81
AK168057.1 - 9 - 10 1564 81
BC006697.1 - 9 - 10 1300 3498573 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000585.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpaa1

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor biosynthetic process
GPI-anchor transamidase activity
GPI anchor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004067.mm.1 NM_010331.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004067.mm.1 NM_010331.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500004067.mm.1 TC1500000585.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0