This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200481_m1
See other SLA GE Assays ›
Gene Symbol:
Sla
Gene Name:
src-like adaptor
Gene Aliases:
Slap, Slap-1
Chromosome Location:
Chr.15: 66780819 - 66831829 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200481_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sla
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029841.4 NP_001025012.1 5 - 6 654 74
NM_009192.3 NP_033218.1 6 - 7 923 74
GenBank mRNA AJ131777.1 - 5 - 6 561 74
AK036167.1 - 5 - 6 655 74
AK037901.1 - 5 - 6 568 74
AK041565.1 - 4 - 5 433 74
AK137954.1 - 5 - 6 589 74
BC032922.1 - 5 - 6 459 1430090 74
CT010282.1 - 4 - 5 357 74
U29056.1 - 4 - 5 375 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001656.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sla

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500012418.mm.1 NM_001029841.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500012418.mm.1 NM_009192.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500012418.mm.1 TC1500001656.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0