This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01199515_g1
See other GNAI3 GE Assays ›
Gene Symbol:
Gnai3
Gene Name:
guanine nucleotide binding protein (G protein), alpha inhibiting 3
Gene Aliases:
AI158965, AW537698, Galphai3, Gnai-3
Chromosome Location:
Chr.3: 108107275 - 108149956 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01199515_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnai3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010306.3 NP_034436.1 2 - 3 304 71
GenBank mRNA AK004566.1 - 2 - 3 298 71
AK077490.1 - 2 - 3 266 71
AK133749.1 - 2 - 3 245 71
AK167438.1 - 2 - 3 258 71
AK168578.1 - 2 - 3 242 71
AK169067.1 - 2 - 3 249 71
AK169285.1 - 2 - 3 279 71
AK187104.1 - 2 - 3 69 71
BC003462.1 - 2 - 3 232 71
BC020009.1 - 2 - 3 276 71
BC041107.1 - 2 - 3 215 2648164 71
BY126665.1 - 2 - 3 305 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002731.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnai3

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

vesicle fusion
cell cycle
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
regulation of heart contraction
cell division
nucleotide binding
GTPase activity
signal transducer activity
protein binding
GTP binding
guanyl nucleotide binding
protein domain specific binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
GTPase activating protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300021417.mm.1 NM_010306.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021417.mm.1 TC0300002731.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0